missing translation for 'onlineSavingsMsg'
Learn More

Serpin G1/C1 Inhibitor Antibody (350507) [DyLight 594], Novus Biologicals™

Product Code. 30734957 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734957 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734957 Supplier Novus Biologicals Supplier No. FAB2488M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Serpin G1/C1 Inhibitor Monoclonal antibody specifically detects Serpin G1/C1 Inhibitor in Human samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serpin G1/C1 Inhibitor
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 350507
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias C1IN, C1INH, C1NH, HAE1, HAE2, SERPING1 serpin peptidase inhibitor, clade G (C1 inhibitor), member 1
Host Species Mouse
Immunogen Human plasma-derived Serpin G1/C1 Inhibitor
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 710
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.