missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin F2/alpha 2-Antiplasmin Rabbit anti-Human, Clone: 25, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | Serpin F2/alpha 2-Antiplasmin |
|---|---|
| Clone | 25 |
| Dilution | Sandwich ELISA Detection 1:1000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18211466
|
Novus Biologicals
NBP2-89396-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18253746
|
Novus Biologicals
NBP2-89396-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serpin F2/alpha 2-Antiplasmin Monoclonal specifically detects Serpin F2/alpha 2-Antiplasmin in Human samples. It is validated for Sandwich ELISA Detection.Specifications
| Serpin F2/alpha 2-Antiplasmin | |
| Sandwich ELISA Detection 1:1000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 5345 | |
| IgG | |
| Protein A purified |
| 25 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| AAPclade F (alpha-2 antiplasmin, Alpha-2-AP, Alpha-2-PI, Alpha-2-plasmin inhibitor, pigment epithelium derived factor), member 2, Serpin F2, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 2 | |
| Recombinant Human Serpin F2/alpha 2-Antiplasmin Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title