missing translation for 'onlineSavingsMsg'
Learn More

Serpin F2/alpha 2-Antiplasmin Antibody (RM0324-2H63), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18405446 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405446 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405446 Supplier Novus Biologicals Supplier No. NBP211890MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Serpin F2/alpha 2-Antiplasmin Monoclonal antibody specifically detects Serpin F2/alpha 2-Antiplasmin in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serpin F2/alpha 2-Antiplasmin
Applications Western Blot
Classification Monoclonal
Clone RM0324-2H63
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias AAPclade F (alpha-2 antiplasmin, Alpha-2-AP, Alpha-2-PI, Alpha-2-plasmin inhibitor, pigment epithelium derived factor), member 2, Serpin F2, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 2
Host Species Rat
Immunogen Mouse recombinant Serpin F2/alpha 2-Antiplasmin
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 5345
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.