missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin F2/alpha 2-Antiplasmin Antibody (RM0324-2H63), Alexa Fluor™ 647, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-11890AF647
This item is not returnable.
View return policy
Description
Serpin F2/alpha 2-Antiplasmin Monoclonal specifically detects Serpin F2/alpha 2-Antiplasmin in Mouse samples. It is validated for Western Blot.
Specifications
| Serpin F2/alpha 2-Antiplasmin | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SERPINF2 | |
| Mouse recombinant Serpin F2/alpha 2-Antiplasmin | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| RM0324-2H63 | |
| Western Blot | |
| AAPclade F (alpha-2 antiplasmin, Alpha-2-AP, Alpha-2-PI, Alpha-2-plasmin inhibitor, pigment epithelium derived factor), member 2, Serpin F2, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 2 | |
| Rat | |
| Protein A or G purified | |
| Cardiovascular Biology | |
| 5345 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction