missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin F2/alpha 2-Antiplasmin Antibody (043), mFluor Violet 610 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89397MFV610
This item is not returnable.
View return policy
Description
Serpin F2/alpha 2-Antiplasmin Monoclonal antibody specifically detects Serpin F2/alpha 2-Antiplasmin in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| Serpin F2/alpha 2-Antiplasmin | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry (Paraffin) | |
| 043 | |
| ELISA, Immunohistochemistry-Paraffin | |
| AAPclade F (alpha-2 antiplasmin, Alpha-2-AP, Alpha-2-PI, Alpha-2-plasmin inhibitor, pigment epithelium derived factor), member 2, Serpin F2, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin F2/alpha 2-Antiplasmin (Accession#: NP_000925.2; Met 1-Lys 491). | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 5345 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction