missing translation for 'onlineSavingsMsg'
Learn More

Serpin F2/alpha 2-Antiplasmin Antibody (001), DyLight 594, Novus Biologicals™

Product Code. 18390400 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390400 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390400 Supplier Novus Biologicals Supplier No. NBP289394DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Serpin F2/alpha 2-Antiplasmin Monoclonal specifically detects Serpin F2/alpha 2-Antiplasmin in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serpin F2/alpha 2-Antiplasmin
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1
Conjugate DyLight 594
Formulation 50 mM Sodium Borate
Gene Alias AAPclade F (alpha-2 antiplasmin, Alpha-2-AP, Alpha-2-PI, Alpha-2-plasmin inhibitor, pigment epithelium derived factor), member 2, Serpin F2, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin F2/alpha 2-Antiplasmin (NP_000925.2; Met 1-Lys 491) .
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 5345
Test Specificity No cross-reactivity in ELISA with: Human SERPINA1 Human SERPINA5 Mouse SERPIND1 Mouse SERPINF1 Mouse SERPINF2
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.