missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin F2/alpha 2-Antiplasmin Antibody (001), Alexa Fluor™ 532, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Serpin F2/alpha 2-Antiplasmin Monoclonal specifically detects Serpin F2/alpha 2-Antiplasmin in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | Serpin F2/alpha 2-Antiplasmin |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 1 |
| Conjugate | Alexa Fluor 532 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | AAPclade F (alpha-2 antiplasmin, Alpha-2-AP, Alpha-2-PI, Alpha-2-plasmin inhibitor, pigment epithelium derived factor), member 2, Serpin F2, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 2 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin F2/alpha 2-Antiplasmin (NP_000925.2; Met 1-Lys 491) . |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?