missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin F1/PEDF Rabbit anti-Human, Clone: 33, Novus Biologicals™
Rabbit Monoclonal Antibody
£299.00 - £443.00
Specifications
| Antigen | Serpin F1/PEDF |
|---|---|
| Clone | 33 |
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18218426
|
Novus Biologicals
NBP2-89799-50UL |
50 μg |
£299.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18243926
|
Novus Biologicals
NBP2-89799-100UL |
100 μg |
£443.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serpin F1/PEDF Monoclonal specifically detects Serpin F1/PEDF in Human samples. It is validated for Western Blot.Specifications
| Serpin F1/PEDF | |
| Western Blot 1:500-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Cell proliferation-inducing gene 35 protein, EPC-1PIG35, PEDFpigment epithelium-derived factor, pigment epithelium derived factor), member 1, proliferation-inducing protein 35, serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, Serpin F1, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 1 | |
| Recombinant Human Serpin F1/PEDF protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 33 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Biologically Active Proteins, Neuroscience, Vision | |
| 0.2 um filtered solution in PBS | |
| 5176 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title