missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin F1/PEDF Antibody (JE41-62), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32733
This item is not returnable.
View return policy
Description
Serpin F1/PEDF Monoclonal antibody specifically detects Serpin F1/PEDF in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| Serpin F1/PEDF | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| Cell proliferation-inducing gene 35 protein, EPC-1PIG35, PEDFpigment epithelium-derived factor, pigment epithelium derived factor), member 1, proliferation-inducing protein 35, serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, Serpin F1, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 1 | |
| Synthetic peptide within Human Serpin F1/PEDF aa 1-50/ 418. (Uniprot: P36955) | |
| 100 μL | |
| Biologically Active Proteins, Neuroscience, Vision | |
| 5176 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE41-62 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction