missing translation for 'onlineSavingsMsg'
Learn More

Serpin F1/PEDF Antibody (187003) [PE], Novus Biologicals™

Product Code. 30735673 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735673 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735673 Supplier Novus Biologicals Supplier No. FAB1177P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Serpin F1/PEDF Monoclonal antibody specifically detects Serpin F1/PEDF in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serpin F1/PEDF
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 187003
Conjugate PE
Formulation PBS
Gene Alias Cell proliferation-inducing gene 35 protein, EPC-1PIG35, PEDFpigment epithelium-derived factor, pigment epithelium derived factor), member 1, proliferation-inducing protein 35, serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, Serpin F1, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 1
Host Species Mouse
Immunogen E. coli-derived recombinant human Serpin F1, Mouse myeloma cell line NS0-derived recombinant human Serpin F1, and S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Serpin F1, Asp44-Pro418, Accession # P36955
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Neuroscience, Vision
Primary or Secondary Primary
Gene ID (Entrez) 5176
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.