missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin F1/PEDF Antibody (033), Alexa Fluor™ 750, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89799AF750
This item is not returnable.
View return policy
Description
Serpin F1/PEDF Monoclonal specifically detects Serpin F1/PEDF in Human samples. It is validated for Western Blot.
Specifications
| Serpin F1/PEDF | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Cell proliferation-inducing gene 35 protein, EPC-1PIG35, PEDFpigment epithelium-derived factor, pigment epithelium derived factor), member 1, proliferation-inducing protein 35, serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, Serpin F1, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin F1/PEDF (NP_002606.3; Met1-Pro418). | |
| 0.1 mL | |
| Biologically Active Proteins, Neuroscience, Vision | |
| 5176 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| 33 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction