missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin E2/PN1 Antibody (OTI2C9), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74117MFV500
This item is not returnable.
View return policy
Description
Serpin E2/PN1 Monoclonal antibody specifically detects Serpin E2/PN1 in Human, Mouse samples. It is validated for Flow Cytometry
Specifications
| Serpin E2/PN1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| OTI2C9 | |
| Flow Cytometry | |
| GDNserine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 2, glia-derived nexin, glial-derived neurite promoting factor, Peptidase inhibitor 7, PI-7, PI7DKFZp686A13110, PN-1, PN1nexin, PNI, Protease nexin 1, Protease nexin I, Serpin E2, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 2 | |
| Full length human recombinant protein of human SERPINE2(NP_006207) produced in HEK293T cell. | |
| 0.1 mL | |
| Neuroscience | |
| 5270 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction