missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin E1/PAI-1 Mouse anti-Human, Clone: 1C5, Novus Biologicals™
Mouse Monoclonal Antibody
£250.00 - £397.00
Specifications
| Antigen | Serpin E1/PAI-1 |
|---|---|
| Clone | 1C5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1086, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18288732
|
Novus Biologicals
NBP2-59495-50UL |
50 μL |
£250.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18246703
|
Novus Biologicals
NBP2-59495-100UL |
100 μL |
£397.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serpin E1/PAI-1 Monoclonal specifically detects Serpin E1/PAI-1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Serpin E1/PAI-1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 5054 | |
| Recombinant human PAI1 (24-402aa) purified from E. coli. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1C5 | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1086, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Endothelial plasminogen activator inhibitor, PAI1PAI-1, PAISerpin E1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1 | |
| SERPINE1 | |
| IgG1 κ | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title