missing translation for 'onlineSavingsMsg'
Learn More

Serpin D1/Heparin Cofactor II Antibody (373008) [FITC], Novus Biologicals™

Product Code. 30734814 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734814 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734814 Supplier Novus Biologicals Supplier No. FAB3198F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Serpin D1/Heparin Cofactor II Monoclonal antibody specifically detects Serpin D1/Heparin Cofactor II in Human samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serpin D1/Heparin Cofactor II
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 373008
Conjugate FITC
Formulation PBS
Gene Alias HC2LS2, HCF2clade D (heparin cofactor), member 1, HC-II, Heparin cofactor II, HLS2HCII, leuserpin 2, Protease inhibitor leuserpin-2, Serpin D1, serpin peptidase inhibitor, clade D (heparin cofactor), member 1
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Serpin D1/Heparin Cofactor II, Gly20-Arg498, Accession # P05546
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 3053
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.