missing translation for 'onlineSavingsMsg'
Learn More

Serpin D1/Heparin Cofactor II Antibody (008), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18331160 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18331160 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18331160 Supplier Novus Biologicals Supplier No. NBP289392AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Serpin D1/Heparin Cofactor II Monoclonal specifically detects Serpin D1/Heparin Cofactor II in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serpin D1/Heparin Cofactor II
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias HC2LS2, HCF2clade D (heparin cofactor), member 1, HC-II, Heparin cofactor II, HLS2HCII, leuserpin 2, Protease inhibitor leuserpin-2, Serpin D1, serpin peptidase inhibitor, clade D (heparin cofactor), member 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin D1/Heparin Cofactor II (NP_000176.2; Met1-Ser499).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 3053
Test Specificity Has cross-reactivity in ELISA with: Mouse SerpinD1 No cross-reactivity in ELISA with: Human SerpinA3 Human SerpinC1 Human SerpinI1 Mouse SerpinF1
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.