missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin C1/Antithrombin-III Antibody (OTI8D5), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74113PECY55
This item is not returnable.
View return policy
Description
Serpin C1/Antithrombin-III Monoclonal specifically detects Serpin C1/Antithrombin-III in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| Serpin C1/Antithrombin-III | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| SERPINC1 | |
| Full length recombinant protein of human Serpin C1/Antithrombin-III (NP_000479) produced in HEK293T cells. | |
| 0.1 mL | |
| 462.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI8D5 | |
| Flow Cytometry | |
| AT3antithrombin-III, ATIIIantithrombin III, MGC22579, serine (or cysteine) proteinase inhibitor, clade C (antithrombin), member 1, serine-cysteine proteinase inhibitor clade C member 1, Serpin C1, serpin peptidase inhibitor, clade C (antithrombin), member 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction