missing translation for 'onlineSavingsMsg'
Learn More

Serpin C1/Antithrombin-III Antibody (023), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18376300 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18376300 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18376300 Supplier Novus Biologicals Supplier No. NBP289296AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Serpin C1/Antithrombin-III Monoclonal specifically detects Serpin C1/Antithrombin-III in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serpin C1/Antithrombin-III
Applications ELISA
Classification Monoclonal
Clone 23
Conjugate Alexa Fluor 647
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias AT3antithrombin-III, ATIIIantithrombin III, MGC22579, serine (or cysteine) proteinase inhibitor, clade C (antithrombin), member 1, serine-cysteine proteinase inhibitor clade C member 1, Serpin C1, serpin peptidase inhibitor, clade C (antithrombin), member 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin C1/Antithrombin-III (NP_000479.1; Met 1-Lys 464).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 462
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.