missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Serpin B8/Proteinase Inhibitor 8 Antibody, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Serpin B8/Proteinase Inhibitor 8 Polyclonal antibody specifically detects Serpin B8/Proteinase Inhibitor 8 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Serpin B8/Proteinase Inhibitor 8 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS (pH 7.4) |
| Gene Accession No. | NP_001027018.1 |
| Gene Alias | CAP-2, CAP2clade B (ovalbumin), member 8, Cytoplasmic antiproteinase 2, Peptidase inhibitor 8, PI-8, protease inhibitor 8 (ovalbumin type), serpin B8, serpin peptidase inhibitor, clade B (ovalbumin), member 8 |
| Host Species | Rabbit |
| Immunogen | SERPINB8 (NP_001027018.1, 1 a.a. - 242 a.a.) full-length human protein. MDDLCEANGTFAISLFKILGEEDNSRNVFFSPMSISSALAMVFMGAKGSTAAQMSQALCLYKDGDIHRGFQSLLSEVNRTGTQYLLRTANRLFGEKTCDFLPDFKEYCQKFYQAELEELSFAEDTEECRKHINDWVAEKTEGKISEVLDAGTVDPLTKLVLVNAIYFKGKWNEQFDRKYTRGMLFKTNEEKKTVQMMFKEAKFKMGYADEVHTQVLELPYVEEELSMVILLPDDNTDLAVKE |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?