missing translation for 'onlineSavingsMsg'
Learn More

Serpin B8/Proteinase Inhibitor 8 Antibody (423023) [Biotin], Novus Biologicals™

Product Code. 30732106 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732106 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732106 Supplier Novus Biologicals Supplier No. FAB4158B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Serpin B8/Proteinase Inhibitor 8 Monoclonal antibody specifically detects Serpin B8/Proteinase Inhibitor 8 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serpin B8/Proteinase Inhibitor 8
Applications Immunohistochemistry
Classification Monoclonal
Clone 423023
Conjugate Biotin
Formulation PBS
Gene Alias CAP-2, CAP2clade B (ovalbumin), member 8, Cytoplasmic antiproteinase 2, Peptidase inhibitor 8, PI-8, protease inhibitor 8 (ovalbumin type), serpin B8, serpin peptidase inhibitor, clade B (ovalbumin), member 8
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf21-derived recombinant human Serpin B8/Proteinase Inhibitor 8, Asp2-Pro374, Accession # P50452
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 5271
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.