missing translation for 'onlineSavingsMsg'
Learn More

Serpin B8/Proteinase Inhibitor 8 Antibody (013), DyLight 405, Novus Biologicals™

Product Code. 18393511 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393511 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393511 Supplier Novus Biologicals Supplier No. NBP290484V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Serpin B8/Proteinase Inhibitor 8 Monoclonal specifically detects Serpin B8/Proteinase Inhibitor 8 in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serpin B8/Proteinase Inhibitor 8
Applications ELISA
Classification Monoclonal
Clone 13
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias CAP-2, CAP2clade B (ovalbumin), member 8, Cytoplasmic antiproteinase 2, Peptidase inhibitor 8, PI-8, protease inhibitor 8 (ovalbumin type), serpin B8, serpin peptidase inhibitor, clade B (ovalbumin), member 8
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Serpin B8/Proteinase Inhibitor 8 (NP_035589.1; Met 1-Pro 374).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 5271
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.