missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin B8/Proteinase Inhibitor 8 Antibody (013), Alexa Fluor™ 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90484AF488
This item is not returnable.
View return policy
Description
Serpin B8/Proteinase Inhibitor 8 Monoclonal specifically detects Serpin B8/Proteinase Inhibitor 8 in Mouse samples. It is validated for ELISA.
Specifications
| Serpin B8/Proteinase Inhibitor 8 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| CAP-2, CAP2clade B (ovalbumin), member 8, Cytoplasmic antiproteinase 2, Peptidase inhibitor 8, PI-8, protease inhibitor 8 (ovalbumin type), serpin B8, serpin peptidase inhibitor, clade B (ovalbumin), member 8 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Serpin B8/Proteinase Inhibitor 8 (NP_035589.1; Met 1-Pro 374). | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 5271 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 13 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction