missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin B6 Antibody (309), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90150CL1
This item is not returnable.
View return policy
Description
Serpin B6 Monoclonal antibody specifically detects Serpin B6 in Human samples. It is validated for ELISA
Specifications
| Serpin B6 | |
| Monoclonal | |
| CoraFluor 1 | |
| CAPMGC111370, Cytoplasmic antiproteinase, deafness, autosomal recessive 91, DFNB91, Peptidase inhibitor 6, PI-6, PI6MSTP057, Placental thrombin inhibitor, protease inhibitor 6 (placental thrombin inhibitor), PTIDKFZp686I04222, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 6, serpin B6, serpin peptidase inhibitor, clade B (ovalbumin), member 6, SPI3 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin B6 (Accession#: AAB30320.1; Asp 2-Pro 376). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 309 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 5269 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction