missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Serpin B6 Monoclonal specifically detects Serpin B6 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | Serpin B6 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 309 |
| Conjugate | Biotin |
| Dilution | ELISA |
| Formulation | PBS |
| Gene Alias | CAPMGC111370, Cytoplasmic antiproteinase, deafness, autosomal recessive 91, DFNB91, Peptidase inhibitor 6, PI-6, PI6MSTP057, Placental thrombin inhibitor, protease inhibitor 6 (placental thrombin inhibitor), PTIDKFZp686I04222, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 6, serpin B6, serpin peptidase inhibitor, clade B (ovalbumin), member 6, SPI3 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin B6 (AAB30320.1; Asp 2-Pro 376). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?