missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin B6 Antibody (202), Novus Biologicals™
Rabbit Monoclonal Antibody
£214.00 - £353.00
Specifications
| Antigen | Serpin B6 |
|---|---|
| Clone | 202 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18629975
|
Novus Biologicals
NBP3-05916-100ul |
100 μg |
£353.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18647175
|
Novus Biologicals
NBP3-05916-50ul |
50 μg |
£214.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serpin B6 Monoclonal antibody specifically detects Serpin B6 in Mouse samples. It is validated for ELISASpecifications
| Serpin B6 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 5269 | |
| IgG | |
| Protein A purified |
| 202 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| CAPMGC111370, Cytoplasmic antiproteinase, deafness, autosomal recessive 91, DFNB91, Peptidase inhibitor 6, PI-6, PI6MSTP057, Placental thrombin inhibitor, protease inhibitor 6 (placental thrombin inhibitor), PTIDKFZp686I04222, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 6, serpin B6, serpin peptidase inhibitor, clade B (ovalbumin), member 6, SPI3 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Serpin B6 (Accession#: NP_035584.1; Met1-Pro377). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title