missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin B5/Maspin Antibody, R&D Systems™
Rat Monoclonal Antibody
£332.60
Specifications
Antigen | Serpin B5/Maspin |
---|---|
Clone | 305339 |
Concentration | LYOPH |
Dilution | Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL, Knockout Validated |
Applications | Western Blot, Immunoprecipitation, Immunoassay |
Description
Serpin B5/Maspin Monoclonal specifically detects Serpin B5/Maspin in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Serpin B5/Maspin | |
LYOPH | |
Western Blot, Immunoprecipitation, Immunoassay | |
Unconjugated | |
Rat | |
Human | |
P36952 | |
5268 | |
E. coli-derived recombinant human Serpin B5 Met1-Pro375 Accession # P36952 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Serpin B5/Maspin in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Serpin A5, B6, B8 or B9 is observed. |
305339 | |
Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL, Knockout Validated | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
maspin, Peptidase inhibitor 5, PI5, PI-5, PI5protease inhibitor 5 (maspin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 5, serpin B5, serpin peptidase inhibitor, clade B (ovalbumin), member 5 | |
SERPINB5 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only