missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin B3/SCCA1 Rabbit anti-Human, Clone: 18, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | Serpin B3/SCCA1 |
|---|---|
| Clone | 18 |
| Dilution | Immunoprecipitation 4-8 ul/mg of lysate |
| Applications | Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18217167
|
Novus Biologicals
NBP2-90195-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18236596
|
Novus Biologicals
NBP2-90195-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serpin B3/SCCA1 Monoclonal specifically detects Serpin B3/SCCA1 in Human samples. It is validated for Immunoprecipitation.Specifications
| Serpin B3/SCCA1 | |
| Immunoprecipitation 4-8 ul/mg of lysate | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| HsT1196, Protein T4-A, SCC, SCCA, SCCA1SCCA-1, SCCA-PD, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3, serpin B3, serpin peptidase inhibitor, clade B (ovalbumin), member 3, Squamous cell carcinoma antigen 1, T4-A | |
| Recombinant Human Serpin B3/SCCA1 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 18 | |
| Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| 0.2 um filtered solution in PBS | |
| 6317 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title