missing translation for 'onlineSavingsMsg'
Learn More

Serpin A9/Centerin Antibody [HRP], Novus Biologicals™

Product Code. 30731506 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731506 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731506 Supplier Novus Biologicals Supplier No. AF5429H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Serpin A9/Centerin Polyclonal antibody specifically detects Serpin A9/Centerin in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serpin A9/Centerin
Applications Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias GCET1, SERPINA11, SERPINA11b
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Serpin A9, Ala24-Thr417, Accession # AAQ89063
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 327657
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.