missing translation for 'onlineSavingsMsg'
Learn More

Serpin A8/Angiotensinogen Antibody, DyLight 650, Novus Biologicals™

Product Code. 18359741 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359741 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18359741 Supplier Novus Biologicals Supplier No. NBP299680C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Serpin A8/Angiotensinogen Polyclonal specifically detects Serpin A8/Angiotensinogen in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serpin A8/Angiotensinogen
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias alpha-1 antiproteinase, antitrypsin, angiotensin II, angiotensinogen, angiotensinogen (serpin peptidase inhibitor, clade A, member 8), ANHU, FLJ92595, FLJ97926, pre-angiotensinogen, serine (or cysteine) proteinase inhibitor, Serpin A8, SERPINA8angiotensin I
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Serpin A8/Angiotensinogen (NP_000020.1; Met 1-Ala 485).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 183
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.