missing translation for 'onlineSavingsMsg'
Learn More

Serpin A8/Angiotensinogen Antibody (1), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18376041 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18376041 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18376041 Supplier Novus Biologicals Supplier No. NBP289736AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Serpin A8/Angiotensinogen Monoclonal specifically detects Serpin A8/Angiotensinogen in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serpin A8/Angiotensinogen
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 1
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias alpha-1 antiproteinase, antitrypsin, angiotensin II, angiotensinogen, angiotensinogen (serpin peptidase inhibitor, clade A, member 8), ANHU, FLJ92595, FLJ97926, pre-angiotensinogen, serine (or cysteine) proteinase inhibitor, Serpin A8, SERPINA8angiotensin I
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin A8/Angiotensinogen (NP_000020.1; Met 1-Ala 485).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 183
Test Specificity No cross-reactivity in ELISA with: Human SerpinA1/ A1AT Human SerpinA3/ SERPINA3/ AACT Human SerpinA5/ Protein C inhibitor Human SerpinD1/ HC2 Human SerpinF2/ SERPINF2 Human SerpinG1/ C1IN Human SerpinI1/ Protease inhibitor 12 Human SerpinC1/SERPINC1/ Antithrombin-III
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.