missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A7/TBG Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99678CL1
This item is not returnable.
View return policy
Description
Serpin A7/TBG Polyclonal antibody specifically detects Serpin A7/TBG in Human samples. It is validated for ELISA
Specifications
| Serpin A7/TBG | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 6906 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| CoraFluor 1 | |
| antitrypsin), member 7, FLJ17820, member 7, Serpin A7, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), T4-binding globulin, TBGclade A (alpha-1 antiproteinase, thyroxin-binding globulin, thyroxine-binding globulin | |
| Produced in rabbits immunized with purified, recombinant Human Serpin A7/TBG (Accession#: NP_000345.2; Met 1-Ala 415) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction