missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A6/Cortisol Binding Globulin Antibody (014), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90529B
This item is not returnable.
View return policy
Description
Serpin A6/Cortisol Binding Globulin Monoclonal specifically detects Serpin A6/Cortisol Binding Globulin in Mouse samples. It is validated for ELISA.
Specifications
| Serpin A6/Cortisol Binding Globulin | |
| Monoclonal | |
| Biotin | |
| antitrypsin), member 6, CBGcorticosteroid-binding globulin, corticosteroid binding globulin, member 6, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A6, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Transcortin | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Serpin A6/Cortisol Binding Globulin (Q06770; Met1-Ala397). | |
| 0.1 mL | |
| Primary | |
| No cross-reactivity in ELISA with: Human SerpinA1 Human SerpinA3 Human SerpinA5 Human Serpinb6 Mouse SerpinB8 Human Serpinb9 Mouse SerpinD1 Mouse SerpinF1 Human SerpinG1 Mouse SerpinI1 Human SerpinI2 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 14 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 866 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction