missing translation for 'onlineSavingsMsg'
Learn More

Serpin A6/Cortisol Binding Globulin Antibody (014), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18377151 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377151 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377151 Supplier Novus Biologicals Supplier No. NBP290529AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Serpin A6/Cortisol Binding Globulin Monoclonal specifically detects Serpin A6/Cortisol Binding Globulin in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serpin A6/Cortisol Binding Globulin
Applications ELISA
Classification Monoclonal
Clone 14
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias antitrypsin), member 6, CBGcorticosteroid-binding globulin, corticosteroid binding globulin, member 6, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A6, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Transcortin
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Serpin A6/Cortisol Binding Globulin (Q06770; Met1-Ala397).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 866
Test Specificity No cross-reactivity in ELISA with: Human SerpinA1 Human SerpinA3 Human SerpinA5 Human Serpinb6 Mouse SerpinB8 Human Serpinb9 Mouse SerpinD1 Mouse SerpinF1 Human SerpinG1 Mouse SerpinI1 Human SerpinI2
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.