missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A5/Protein C Inhibitor Antibody (210326) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1266JF635
This item is not returnable.
View return policy
Description
Serpin A5/Protein C Inhibitor Monoclonal antibody specifically detects Serpin A5/Protein C Inhibitor in Human samples. It is validated for Immunoprecipitation,Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| Serpin A5/Protein C Inhibitor | |
| Monoclonal | |
| Janelia Fluor 635 | |
| Acrosomal serine protease inhibitor, antitrypsin), member 5, member 5, PAI-3, PAI3PLANH3, PCIplasminogen activator inhibitor III, Plasminogen activator inhibitor 3, plasminogen activator inhibitor-3, PROCIplasma serine protease inhibitor, Protein C inhibitor, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A5, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), SERPNA5 | |
| Mouse myeloma cell line NS0-derived recombinant human Serpin A5/Protein C, His20-Pro406, Accession # P05154 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Immunoprecipitation, Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 210326 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.5 mL | |
| Cancer | |
| 5104 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction