missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Serpin A4 ELISA Kit

Product Code. 15475123
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
1 set
Each
This item is not returnable. View return policy

Product Code. 15475123

Brand: Invitrogen™ EHSERPINA4

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The Human Serpin A4 ELISA quantitates Hu Serpin A4 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Serpin A4. Principle of the method The Human Serpin A4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

SERPINA4 inhibits human amidolytic and kininogenase activities of tissue kallikrein. The inhibition is achieved by formation of an equimolar, heat- and SDS-stable complex between the inhibitor and the enzyme, and generation of a small C-terminal fragment of the inhibitor due to cleavage at the reactive site by tissue kallikrein.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P29622
Assay Range 0.143 to 35 ng/mL
Assay Sensitivity 0.12 ng/mL
Calibration Range 0.12 ng/mL to 35 ng/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Product Type ELISA
Protein Family Proteases, Peptidases & Inhibitors
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA pair antibody detects human Serpin A4. Other species not determined.
Sufficient For 96 Assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 5267
Gene Symbol KAL,KLST,KST,PI-4,PI4,kallistatin
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Other Research
Sample Volume Plasma 1 μL, Serum 1 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Synonym KAL, KLST, KST, PI-4, PI4, kallistatin
Target Kallistatin
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.