missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Serpin A3/alpha 1-Antichymotrypsin Antibody Pair

Product Code. 18654510 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18654510

Brand: Novus Biologicals™ H00000012AP11

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Assay Range 3 ng/mL to 100 ng/mL
Conjugate Unconjugated
Product Type Sandwich ELISA
For Use With (Application) Sandwich ELISA
Gene ID (Entrez) 12
Gene Symbol SERPINA3
Quantity 1 Set
Research Discipline Breast Cancer, Cancer
Storage Requirements Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Synonym AACTMGC88254, ACTGIG25, alpha-1-antichymotrypsin, antitrypsin), member 3, Cell growth-inhibiting gene 24/25 protein, growth-inhibiting protein 24, growth-inhibiting protein 25, member 3, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 3, Serpin A3, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin)
Target Human
Buffer 1x PBS, pH 7.4
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.