Learn More
Serpin A3/alpha 1-Antichymotrypsin Antibody (CL10697) [Alexa Fluor™ 750], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-07987AF750
Product Code. 30292502
  • £558.00 / 0.10mL
Description
Serpin A3/alpha 1-Antichymotrypsin Monoclonal antibody specifically detects Serpin A3/alpha 1-Antichymotrypsin in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Serpin A3/alpha 1-Antichymotrypsin
Monoclonal
Alexa Fluor 750
AACTMGC88254, ACTGIG25, alpha-1-antichymotrypsin, antitrypsin), member 3, Cell growth-inhibiting gene 24/25 protein, growth-inhibiting protein 24, growth-inhibiting protein 25, member 3, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 3, Serpin A3, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin)
This antibody was developed using a recombinant protein derived from P01011, with the exact immunogen sequence remaining proprietary.
0.1 mL
Breast Cancer, Cancer
12
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL10697
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified