missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A3/alpha 1-Antichymotrypsin Antibody (AACT/1451), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54339AF700
This item is not returnable.
View return policy
Description
Serpin A3/alpha 1-Antichymotrypsin Monoclonal antibody specifically detects Serpin A3/alpha 1-Antichymotrypsin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Specifications
| Serpin A3/alpha 1-Antichymotrypsin | |
| Monoclonal | |
| Alexa Fluor 700 | |
| AACTMGC88254, ACTGIG25, alpha-1-antichymotrypsin, antitrypsin), member 3, Cell growth-inhibiting gene 24/25 protein, growth-inhibiting protein 24, growth-inhibiting protein 25, member 3, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 3, Serpin A3, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin) | |
| Recombinant human Serpin A3/alpha 1-Antichymotrypsin protein fragment (around aa 49-187) (exact sequence is proprietary) (Uniprot: P01011) | |
| 100 μg | |
| Breast Cancer, Cancer | |
| 12 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| AACT/1451 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction