missing translation for 'onlineSavingsMsg'
Learn More

Serpin A3/alpha 1-Antichymotrypsin Antibody (011), Alexa Fluor™ 700 , Novus Biologicals™

Product Code. 18421837 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421837 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421837 Supplier Novus Biologicals Supplier No. NBP289405AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Serpin A3/alpha 1-Antichymotrypsin Monoclonal antibody specifically detects Serpin A3/alpha 1-Antichymotrypsin in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serpin A3/alpha 1-Antichymotrypsin
Applications ELISA
Classification Monoclonal
Clone 011
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias AACTMGC88254, ACTGIG25, alpha-1-antichymotrypsin, antitrypsin), member 3, Cell growth-inhibiting gene 24/25 protein, growth-inhibiting protein 24, growth-inhibiting protein 25, member 3, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 3, Serpin A3, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin)
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin A3/alpha 1-Antichymotrypsin (NP_001076.2; Met1-Ala423).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 12
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.