missing translation for 'onlineSavingsMsg'
Learn More

Serpin A1a Antibody (882213) [DyLight 405], Novus Biologicals™

Product Code. 30734732 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734732 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734732 Supplier Novus Biologicals Supplier No. FAB7690E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Serpin A1a Monoclonal antibody specifically detects Serpin A1a in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serpin A1a
Applications Western Blot
Classification Monoclonal
Clone 882213
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias A1A, A1AT, AAT, alpha1AT, nNIF, PI, PI1, PRO2275
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Serpin A1a, Met1-Lys413, Accession # P07758
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5265
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.