missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A12 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | Serpin A12 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Descripción
Serpin A12 Polyclonal specifically detects Serpin A12 in Mouse samples. It is validated for Western Blot.Especificaciones
| Serpin A12 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| antitrypsin), member 12, member 12, OL-64Visceral adipose-specific serpin, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serpin A12, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Vaspin, Visceral adipose tissue-derived serine protease inhibitor | |
| The immunogen is a synthetic peptide directed towards the middle region of mouse Serpin A12 (NP_080811.1). Peptide sequence SRLGISKIFEENGDLTRISSHRSLKVGEAVHKAELKMDEKGMEGAAGSGA | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 145264 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto