missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A12 Antibody (VP63), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80041F
This item is not returnable.
View return policy
Description
Serpin A12 Monoclonal antibody specifically detects Serpin A12 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
| Serpin A12 | |
| Monoclonal | |
| FITC | |
| antitrypsin), member 12, member 12, OL-64Visceral adipose-specific serpin, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serpin A12, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Vaspin, Visceral adipose tissue-derived serine protease inhibitor | |
| Recombinant human Serpin A12. | |
| 0.1 mL | |
| metabolism | |
| 145264 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| VP63 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction