missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A12 Antibody (VP63), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-80041C
This item is not returnable.
View return policy
Description
Serpin A12 Monoclonal antibody specifically detects Serpin A12 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
| Serpin A12 | |
| Monoclonal | |
| DyLight 650 | |
| antitrypsin), member 12, member 12, OL-64Visceral adipose-specific serpin, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serpin A12, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Vaspin, Visceral adipose tissue-derived serine protease inhibitor | |
| Recombinant human Serpin A12. | |
| 0.1 mL | |
| metabolism | |
| 145264 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| VP63 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction