missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A12 Antibody (1C4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04354
This item is not returnable.
View return policy
Description
Serpin A12 Monoclonal specifically detects Serpin A12 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Serpin A12 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:3000, ELISA 1:100-1:2000 | |
| antitrypsin), member 12, member 12, OL-64Visceral adipose-specific serpin, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serpin A12, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Vaspin, Visceral adipose tissue-derived serine protease inhibitor | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human Vaspin mAb, clone 1C4, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Vaspin protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 1C4 | |
| Unconjugated | |
| Q8IW75 | |
| SERPINA12 | |
| Recombinant human Vaspin (21-414aa) purified from E. coli (NM_173850). | |
| 0.1 mL | |
| metabolism | |
| 145264 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction