missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A12 Antibody (1C4), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04354AF594
This item is not returnable.
View return policy
Description
Serpin A12 Monoclonal specifically detects Serpin A12 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Serpin A12 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SERPINA12 | |
| Recombinant human Vaspin (21-414aa) purified from E. coli (NM_173850). | |
| metabolism | |
| 145264 | |
| Store at 4°C in the dark. |
| Western Blot, ELISA | |
| 1C4 | |
| Western Blot, ELISA | |
| antitrypsin), member 12, member 12, OL-64Visceral adipose-specific serpin, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serpin A12, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Vaspin, Visceral adipose tissue-derived serine protease inhibitor | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction