missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Serpin A11 Polyclonal specifically detects Serpin A11 in Mouse samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | Serpin A11 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | DyLight 405 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | antiproteinase-like 2, antitrypsin), member 11, member 11, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serpin A11, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin) |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Mouse Serpin A11 (NP_955018.2; Met 1-Gly 424). |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?