missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Serpin A1/alpha 1-Antitrypsin Antibody Pair [HRP]

Product Code. 18247297 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18247297

Brand: Novus Biologicals™ NBP27956515PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Serpin A1/alpha 1-Antitrypsin.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.25 pg/mL
Assay Sensitivity 31.25-2000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Serpin A1/alpha 1-Antitrypsin.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias A1A, AATMGC23330, Alpha-1 protease inhibitor, Alpha-1-antiproteinase, alpha-1-antitrypsin, antitrypsin), member 1, member 1, PIMGC9222, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 1, Serpin A1, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin)
Gene ID (Entrez) 5265
Gene Symbol SERPINA1
Host Species Rabbit/Mouse
Kit Contents
  • Detection Antibody: 0.2 mg/mL rabbit anti-Serpin A1/alpha 1-Antitrypsin monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4).
  • Capture Antibody: 0.5 mg/mL of mouse anti-Serpin A1/alpha 1-Antitrypsin monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 160 ng of recombinant Serpin A1/alpha 1-Antitrypsin.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Stem Cell Markers
    Storage Requirements Storage is content dependent.
    Target Serpin A1/alpha 1-Antitrypsin
    Technique 1. Capture Antibody: Dilute to a working concentration of 1 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.4 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2 ng/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.