missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A1/alpha 1-Antitrypsin Antibody (AAT/1379), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54442H
This item is not returnable.
View return policy
Description
Serpin A1/alpha 1-Antitrypsin Monoclonal antibody specifically detects Serpin A1/alpha 1-Antitrypsin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Specifications
| Serpin A1/alpha 1-Antitrypsin | |
| Monoclonal | |
| HRP | |
| A1A, AATMGC23330, Alpha-1 protease inhibitor, Alpha-1-antiproteinase, alpha-1-antitrypsin, antitrypsin), member 1, member 1, PIMGC9222, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 1, Serpin A1, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin) | |
| Recombinant human Serpin A1/alpha 1-Antitrypsin protein fragment (Uniprot: P01009) | |
| 100 μg | |
| Stem Cell Markers | |
| 5265 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| AAT/1379 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction