missing translation for 'onlineSavingsMsg'
Learn More

Serpin A1/alpha 1-Antitrypsin Antibody (5D1D2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication

£186.00 - £450.00

Specifications

Antigen Serpin A1/alpha 1-Antitrypsin
Clone 5D1D2
Concentration 1 mg/mL
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, Immunofluorescence, CyTOF-ready
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18071704
View Documents
Novus Biologicals
NBP2-52557
0.1 mg
£450.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18650367
View Documents
Novus Biologicals
NBP2-52557-0.025mg
0.025 mg
£186.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Serpin A1/alpha 1-Antitrypsin Monoclonal specifically detects Serpin A1/alpha 1-Antitrypsin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications

Specifications

Serpin A1/alpha 1-Antitrypsin
1 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Stem Cell Markers
PBS with 0.05% Sodium Azide
5265
Purified recombinant fragment of human SERPINA1 (AA: 23-237) expressed in E. Coli.
Primary
46.7 kDa
5D1D2
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, Immunofluorescence, CyTOF-ready
Monoclonal
Purified
RUO
Human
A1A, AATMGC23330, Alpha-1 protease inhibitor, Alpha-1-antiproteinase, alpha-1-antitrypsin, antitrypsin), member 1, member 1, PIMGC9222, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 1, Serpin A1, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin)
SERPINA1
IgG1
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.