missing translation for 'onlineSavingsMsg'
Learn More

Serotonin Hydrochloride, TRC

Product Code. 30010589
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
Unit Size:
100mg
1g
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010589 1 g 1g
17994165 25 mg 25mg
17903355 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010589 Supplier Toronto Research Chemicals Supplier No. TRCS2749801G
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153-98-0
Molecular Formula C10 H12 N2 O . Cl H
Molecular Weight (g/mol) 212.68
Synonym 1H-Indol-5-ol, 3-(2-aminoethyl)-, hydrochloride (1:1), 1H-Indol-5-ol, 3-(2-aminoethyl)-, monohydrochloride (9CI), Indol-5-ol, 3-(2-aminoethyl)-, monohydrochloride (8CI), 5-HT.bul.HCl, 5-Hydroxytryptamine hydrochloride, Hippophaine hydrochloride, Serotonin hydrochloride, 3-(2-Aminoethyl)-1H-indol-5-ol hydrochloride
IUPAC Name 3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride
SMILES Cl.NCCc1c[nH]c2ccc(O)cc12

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H12N2O.ClH/c11-4-3-7-6-12-10-2-1-8(13)5-9(7)10;/h1-2,5-6,12-13H,3-4,11H2;1H
Formula Weight 212.0716
Chemical Name or Material Serotonin hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.