missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Serine racemase Recombinant Rabbit Monoclonal Antibody (K01_2P44)

Product Code. 30828918
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828918 50 μL
30829318 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828918 Supplier Invitrogen™ Supplier No. MA565664
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Serine racemase Recombinant Monoclonal Antibody for Western Blot

SRR (serine racemase) catalyzes the synthesis of D-serine from L-serine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serine racemase
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2P44
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene Srr
Gene Accession No. Q9GZT4
Gene Alias D-serine ammonia-lyase; D-serine dehydratase; ILV1; ISO1; L-serine ammonia-lyase; L-serine dehydratase; Serine racemase; SRR; Srs
Gene Symbols Srr
Host Species Rabbit
Immunogen A synthetic peptide of human Serine racemase.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63826
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.